Assay Details
| Gene Symbol: NEFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646887.1 | XP_023502655.1 | 959 | 109 | ||
| XM_023646888.1 | XP_023502656.1 | 959 | 109 | |||
| XM_023646889.1 | XP_023502657.1 | 959 | 109 | |||
| XM_023646890.1 | XP_023502658.1 | 959 | 109 | |||
| XM_023646891.1 | XP_023502659.1 | 959 | 109 | |||
| XM_023646892.1 | XP_023502660.1 | 959 | 109 | |||
| XM_023646893.1 | XP_023502661.1 | 959 | 109 | |||
| XM_023646894.1 | XP_023502662.1 | 959 | 109 | |||
| XM_023646895.1 | XP_023502663.1 | 958 | 109 | |||
More Information
Set Membership: |
|

NEFH
Genomic Map