Assay Details
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489039.6 | XP_001489089.1 | 125 | 60 | ||
| XM_014742183.2 | XP_014597669.1 | 100 | 60 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489039.6 | XP_001489089.1 | 125 | 60 | ||
| XM_014742183.2 | XP_014597669.1 | 100 | 60 | |||
Set Membership: |
|