Assay Details
| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014727566.2 | XP_014583052.2 | 809 | 58 | ||
| XM_023647042.1 | XP_023502810.1 | 812 | 58 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BICDL1| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014727566.2 | XP_014583052.2 | 809 | 58 | ||
| XM_023647042.1 | XP_023502810.1 | 812 | 58 | |||
Set Membership: |
|