Assay Details
| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005612561.3 | XP_005612618.2 | 972 | 107 | ||
| XM_014727566.2 | XP_014583052.2 | 958 | 107 | |||
| XM_014727567.2 | XP_014583053.2 | 973 | 107 | |||
| XM_023647041.1 | XP_023502809.1 | 974 | 107 | |||
| XM_023647042.1 | XP_023502810.1 | 961 | 107 | |||
More Information
Set Membership: |
|

BICDL1
Genomic Map