Assay Details
| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005612561.3 | XP_005612618.2 | 1514 | 102 | ||
| XM_014727566.2 | XP_014583052.2 | 1500 | 102 | |||
| XM_014727567.2 | XP_014583053.2 | 1515 | 102 | |||
| XM_023647043.1 | XP_023502811.1 | 1335 | 102 | |||
More Information
Set Membership: |
|

BICDL1
Genomic Map