Assay Details
| Gene Symbol: LOC100055035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647059.1 | XP_023502827.1 | 1408 | 77 | ||
| XM_023647060.1 | XP_023502828.1 | 1408 | 77 | |||
| XM_023647061.1 | XP_023502829.1 | 1402 | 77 | |||
More Information
Set Membership: |
|

LOC100055035
Genomic Map