Assay Details
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647210.1 | XP_023502978.1 | 478 | 60 | ||
| XM_023647211.1 | XP_023502979.1 | 467 | 60 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCL7A| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647210.1 | XP_023502978.1 | 478 | 60 | ||
| XM_023647211.1 | XP_023502979.1 | 467 | 60 | |||
Set Membership: |
|