Assay Details
| Gene Symbol: C8H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498367.5 | XP_001498417.2 | 259 | 105 | ||
| XM_003365544.4 | XP_003365592.1 | 264 | 105 | |||
| XM_005612875.3 | XP_005612932.1 | 266 | 105 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001498367.5, XM_003365544.4, XM_005612875.3 transcripts.
Set Membership: |
|

C8H18orf25
Genomic Map