Assay Details
| Gene Symbol: C8H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498367.5 | XP_001498417.2 | 1044 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8H18orf25| Gene Symbol: C8H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498367.5 | XP_001498417.2 | 1044 | 73 | ||