Assay Details
| Gene Symbol: ZBTB7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647771.1 | XP_023503539.1 | 345 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023647771.1 transcripts.
Set Membership: |
|

ZBTB7C
Genomic Map