Assay Details
| Gene Symbol: SERPINB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492201.4 | XP_001492251.1 | 692 | 145 | ||
| XM_014727840.2 | XP_014583326.1 | 590 | 145 | |||
| XM_023647953.1 | XP_023503721.1 | 873 | 145 | |||
| XM_023647954.1 | XP_023503722.1 | 1741 | 145 | |||
More Information
Set Membership: |
|

SERPINB8
Genomic Map