Assay Details
| Gene Symbol: CA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014727918.2 | XP_014583404.2 | 306 | 82 | ||
| XM_014727920.2 | XP_014583406.2 | 393 | 82 | |||
| XM_014727921.2 | XP_014583407.2 | 382 | 82 | |||
| XM_023648370.1 | XP_023504138.1 | 538 | 82 | |||
| XM_023648371.1 | XP_023504139.1 | 540 | 82 | |||
| XM_023648372.1 | XP_023504140.1 | 539 | 82 | |||
More Information
Set Membership: |
|

CA13
Genomic Map