Assay Details
| Gene Symbol: LOC100052030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648506.1 | XP_023504274.1 | 1522 | 92 | ||
| XM_023648507.1 | XP_023504275.1 | 1521 | 92 | |||
| XM_023648508.1 | XP_023504276.1 | 1720 | 92 | |||
| XM_023648509.1 | XP_023504277.1 | 1534 | 92 | |||
| XM_023648510.1 | XP_023504278.1 | 1483 | 92 | |||
More Information
Set Membership: |
|

LOC100052030
Genomic Map