Assay Details
| Gene Symbol: LOC100052030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648506.1 | XP_023504274.1 | 424 | 79 | ||
| XM_023648507.1 | XP_023504275.1 | 423 | 79 | |||
| XM_023648508.1 | XP_023504276.1 | 622 | 79 | |||
| XM_023648509.1 | XP_023504277.1 | 436 | 79 | |||
| XM_023648510.1 | XP_023504278.1 | 424 | 79 | |||
More Information
Set Membership: |
|

LOC100052030
Genomic Map