Assay Details
| Gene Symbol: CIBAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648687.1 | XP_023504455.1 | 472 | 80 | ||
| XM_023648688.1 | XP_023504456.1 | 472 | 80 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIBAR1| Gene Symbol: CIBAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648687.1 | XP_023504455.1 | 472 | 80 | ||
| XM_023648688.1 | XP_023504456.1 | 472 | 80 | |||
Set Membership: |
|