Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489304.5 | XP_001489354.1 | 2218 | 94 | ||
| XM_014728200.2 | XP_014583686.1 | 2218 | 94 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VIRMA| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489304.5 | XP_001489354.1 | 2218 | 94 | ||
| XM_014728200.2 | XP_014583686.1 | 2218 | 94 | |||