Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001491150.6 | XP_001491200.1 | 401 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001491150.6 transcripts.
Set Membership: |
|

RPL30
Genomic Map