Assay Details
| Gene Symbol: ZNF706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003365653.4 | XP_003365701.1 | 424 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF706| Gene Symbol: ZNF706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003365653.4 | XP_003365701.1 | 424 | 86 | ||