Assay Details
Gene Symbol: LY6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649070.1 | XP_023504838.1 | 509 | 65 | ||
XM_023649071.1 | XP_023504839.1 | 437 | 65 | |||
XM_023649072.1 | XP_023504840.1 | 501 | 65 | |||
XM_023649073.1 | XP_023504841.1 | 400 | 65 | |||
XM_023649074.1 | XP_023504842.1 | 561 | 65 | |||
XM_023649075.1 | XP_023504843.1 | 554 | 65 | |||
XM_023649076.1 | XP_023504844.1 | 438 | 65 | |||
XM_023649077.1 | XP_023504845.1 | 601 | 65 | |||
XM_023649078.1 | XP_023504846.1 | 439 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
