Assay Details
| Gene Symbol: C9H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649330.1 | XP_023505098.1 | 3740 | 74 | ||
| XM_023649331.1 | XP_023505099.1 | 5803 | 74 | |||
| XM_023649332.1 | XP_023505100.1 | 5720 | 74 | |||
| XM_023649333.1 | XP_023505101.1 | 1741 | 74 | |||
| XM_023649334.1 | XP_023505102.1 | 1680 | 74 | |||
More Information
Set Membership: |
|

C9H8orf33
Genomic Map