Assay Details
| Gene Symbol: CA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646651.1 | XP_023502419.1 | 1846 | 113 | ||
| XM_023646652.1 | XP_023502420.1 | 1846 | 113 | |||
More Information
Set Membership: |
|

CA15
Genomic Map