Assay Details
| Gene Symbol: CA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646651.1 | XP_023502419.1 | 1911 | 72 | ||
| XM_023646652.1 | XP_023502420.1 | 1911 | 72 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646651.1 | XP_023502419.1 | 1911 | 72 | ||
| XM_023646652.1 | XP_023502420.1 | 1911 | 72 | |||
Set Membership: |
|