Assay Details
| Gene Symbol: LOC100050480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001487926.4 | XP_001487976.2 | 957 | 116 | ||
| XM_023646617.1 | XP_023502385.1 | 796 | 116 | |||
| XM_023646618.1 | XP_023502386.1 | 1088 | 116 | |||
More Information
Set Membership: |
|

LOC100050480
Genomic Map