Assay Details
| Gene Symbol: ABRACL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003365731.4 | XP_003365779.1 | 200 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003365731.4 transcripts.
Set Membership: |
|

ABRACL
Genomic Map