Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640125.1 | XP_023495893.1 | 2572 | 82 | ||
| XM_023640126.1 | XP_023495894.1 | 2472 | 82 | |||
| XM_023640127.1 | XP_023495895.1 | 4067 | 82 | |||
| XM_023640128.1 | XP_023495896.1 | 2047 | 82 | |||
| XM_023640129.1 | XP_023495897.1 | 1169 | 82 | |||
| XM_023640130.1 | XP_023495898.1 | 2728 | 82 | |||
| XM_023640131.1 | XP_023495899.1 | 1260 | 82 | |||
More Information
Set Membership: |
|

DYNC2I1
Genomic Map