Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640125.1 | XP_023495893.1 | 2864 | 79 | ||
| XM_023640126.1 | XP_023495894.1 | 2764 | 79 | |||
| XM_023640127.1 | XP_023495895.1 | 4359 | 79 | |||
| XM_023640128.1 | XP_023495896.1 | 2339 | 79 | |||
| XM_023640129.1 | XP_023495897.1 | 1461 | 79 | |||
| XM_023640130.1 | XP_023495898.1 | 3020 | 79 | |||
| XM_023640131.1 | XP_023495899.1 | 1552 | 79 | |||
More Information
Set Membership: |
|

DYNC2I1
Genomic Map