Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640125.1 | XP_023495893.1 | 199 | 83 | ||
| XM_023640126.1 | XP_023495894.1 | 191 | 83 | |||
| XM_023640127.1 | XP_023495895.1 | 1786 | 83 | |||
More Information
Set Membership: |
|

DYNC2I1
Genomic Map