Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640125.1 | XP_023495893.1 | 1219 | 100 | ||
| XM_023640126.1 | XP_023495894.1 | 1119 | 100 | |||
| XM_023640127.1 | XP_023495895.1 | 2714 | 100 | |||
| XM_023640128.1 | XP_023495896.1 | 694 | 100 | |||
| XM_023640130.1 | XP_023495898.1 | 1289 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023640130.1 transcripts.
Set Membership: |
|

DYNC2I1
Genomic Map