Assay Details
| Gene Symbol: BRME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613572.3 | XP_005613629.1 | 200 | 71 | ||
| XM_005613575.3 | XP_005613632.1 | 269 | 71 | |||
| XM_023645258.1 | XP_023501026.1 | 201 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005613575.3, XM_023645258.1 transcripts.
Set Membership: |
|

BRME1
Genomic Map