Assay Details
| Gene Symbol: BRME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613571.3 | XP_005613628.1 | 465 | 60 | ||
| XM_005613572.3 | XP_005613629.1 | 462 | 60 | |||
| XM_005613573.3 | XP_005613630.1 | 467 | 60 | |||
| XM_005613574.3 | XP_005613631.1 | 448 | 60 | |||
| XM_005613575.3 | XP_005613632.1 | 531 | 60 | |||
| XM_023645257.1 | XP_023501025.1 | 423 | 60 | |||
| XM_023645258.1 | XP_023501026.1 | 463 | 60 | |||
More Information
Set Membership: |
|

BRME1
Genomic Map