Assay Details
| Gene Symbol: BRME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613571.3 | XP_005613628.1 | 567 | 92 | ||
| XM_005613572.3 | XP_005613629.1 | 564 | 92 | |||
| XM_005613573.3 | XP_005613630.1 | 569 | 92 | |||
| XM_005613574.3 | XP_005613631.1 | 550 | 92 | |||
| XM_005613575.3 | XP_005613632.1 | 633 | 92 | |||
| XM_023645257.1 | XP_023501025.1 | 525 | 92 | |||
| XM_023645258.1 | XP_023501026.1 | 565 | 92 | |||

BRME1
Genomic Map