Assay Details
| Gene Symbol: CITED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488044.6 | XP_001488094.1 | 510 | 77 | ||
| XM_003365810.4 | XP_003365858.1 | 341 | 77 | |||
| XM_005614267.3 | XP_005614324.1 | 474 | 77 | |||
| XM_014728966.2 | XP_014584452.1 | 174 | 77 | |||
| XM_023633497.1 | XP_023489265.1 | 369 | 77 | |||
| XM_023633499.1 | XP_023489267.1 | 204 | 77 | |||
| XM_023633500.1 | XP_023489268.1 | 548 | 77 | |||
| XM_023633501.1 | XP_023489269.1 | 200 | 77 | |||
| XM_023633502.1 | XP_023489270.1 | 147 | 77 | |||
More Information
Set Membership: |
|

CITED1
Genomic Map