Assay Details
| Gene Symbol: CYSLTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614289.3 | XP_005614346.1 | 485 | 102 | ||
| GenBank mRNA | JL616299.2 | - | 1 - 2 | 130 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005614289.3 transcripts.
Set Membership: |
|

CYSLTR1
Genomic Map