Assay Details
| Gene Symbol: BEX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003365846.4 | XP_003365894.1 | 241 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003365846.4 transcripts.
Set Membership: |
|

BEX3
Genomic Map