Assay Details
| Gene Symbol: LOC102148739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729077.2 | XP_014584563.1 | 249 | 84 | ||
| XM_023633958.1 | XP_023489726.1 | 251 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014729077.2, XM_023633958.1 transcripts.
Set Membership: |
|

LOC102148739
Genomic Map