Assay Details
| Gene Symbol: LOC102148739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729075.2 | XP_014584561.1 | 368 | 95 | ||
| XM_014729077.2 | XP_014584563.1 | 324 | 95 | |||
| XM_014729078.2 | XP_014584564.1 | 147 | 95 | |||

LOC102148739
Genomic Map