Assay Details
| Gene Symbol: LOC102148739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729075.2 | XP_014584561.1 | 2325 | 92 | ||
| XM_014729077.2 | XP_014584563.1 | 2281 | 92 | |||
| XM_014729078.2 | XP_014584564.1 | 2104 | 92 | |||
| XM_023633957.1 | XP_023489725.1 | 2281 | 92 | |||
| XM_023633958.1 | XP_023489726.1 | 2238 | 92 | |||
| XM_023633959.1 | XP_023489727.1 | 2059 | 92 | |||

LOC102148739
Genomic Map