Assay Details
| Gene Symbol: PABIR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023634397.1 | XP_023490165.1 | 1607 | 75 | ||
| XM_023634398.1 | XP_023490166.1 | 1605 | 75 | |||
| XM_023634399.1 | XP_023490167.1 | 1594 | 75 | |||
| XM_023634400.1 | XP_023490168.1 | 1591 | 75 | |||
| XM_023634401.1 | XP_023490169.1 | 1609 | 75 | |||
| XM_023634402.1 | XP_023490170.1 | 1607 | 75 | |||
| XM_023634403.1 | XP_023490171.1 | 1595 | 75 | |||
| XM_023634404.1 | XP_023490172.1 | 1593 | 75 | |||
More Information
Set Membership: |
|

PABIR2
Genomic Map