Assay Details
Gene Symbol: PABIR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614514.3 | XP_005614571.1 | 886 | 118 | ||
XM_005614516.3 | XP_005614573.1 | 825 | 118 | |||
XM_014729199.2 | XP_014584685.1 | 882 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
