Assay Details
Gene Symbol: PABIR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614514.3 | XP_005614571.1 | 415 | 104 | ||
XM_005614515.3 | XP_005614572.1 | 415 | 104 | |||
XM_005614516.3 | XP_005614573.1 | 354 | 104 | |||
XM_014729197.2 | XP_014584683.1 | 415 | 104 | |||
XM_014729199.2 | XP_014584685.1 | 411 | 104 | |||
GenBank mRNA | JL636786.1 | - | 7 - 8 | 473 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
