Assay Details
| Gene Symbol: LOC100064632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495475.5 | XP_001495525.5 | 126 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001495475.5 transcripts.
Set Membership: |
|

LOC100064632
Genomic Map