Assay Details
| Gene Symbol: LOC100058594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023633522.1 | XP_023489290.1 | 1343 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100058594| Gene Symbol: LOC100058594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023633522.1 | XP_023489290.1 | 1343 | 89 | ||