Assay Details
| Gene Symbol: SQOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618171.1 | XP_023473939.1 | 804 | 85 | ||
| GenBank mRNA | HQ889913.1 | - | 1 - 2 | 133 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SQOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618171.1 | XP_023473939.1 | 804 | 85 | ||
| GenBank mRNA | HQ889913.1 | - | 1 - 2 | 133 | 85 | |
Set Membership: |
|