Assay Details
| Gene Symbol: SQOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618171.1 | XP_023473939.1 | 1197 | 96 | ||
| GenBank mRNA | HQ889913.1 | - | 3 - 4 | 526 | 96 | |
| JL622964.1 | - | 3 - 4 | 564 | 96 | ||
More Information
Set Membership: |
|

SQOR
Genomic Map