Assay Details
| Gene Symbol: TRIAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489061.5 | XP_001489111.2 | 187 | 70 | ||
| GenBank mRNA | JL616119.2 | - | 1 - 2 | 159 | 70 | |
More Information
Set Membership: |
|

TRIAP1
Genomic Map