Assay Details
| Gene Symbol: LAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609575.3 | XP_005609632.1 | 2766 | 98 | ||
| XM_014739444.2 | XP_014594930.1 | 1604 | 98 | |||
| XM_023640318.1 | XP_023496086.1 | 2995 | 98 | |||
| GenBank mRNA | JL621911.1 | - | 1 | 420 | 98 | |
More Information
Set Membership: |
|

LAX1
Genomic Map