Assay Details
| Gene Symbol: TENT5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610285.3 | XP_005610342.1 | 1522 | 81 | ||
| XM_023641335.1 | XP_023497103.1 | 1641 | 81 | |||
| GenBank mRNA | JL624670.1 | - | 1 | 519 | 81 | |
More Information
Set Membership: |
|

TENT5C
Genomic Map