Assay Details
| Gene Symbol: C18H2orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732343.2 | XP_014587829.1 | 1033 | 67 | ||
| GenBank mRNA | JL628518.1 | - | 1 | 415 | 67 | |
More Information
Set Membership: |
|

C18H2orf66
Genomic Map