Assay Details
| Gene Symbol: RPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614325.3 | XP_005614382.1 | 1432 | 136 | ||
| GenBank mRNA | JL636687.1 | - | 1 | 119 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614325.3 | XP_005614382.1 | 1432 | 136 | ||
| GenBank mRNA | JL636687.1 | - | 1 | 119 | 137 | |
Set Membership: |
|