Assay Details
Gene Symbol: OXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625933.1 | XP_023481701.1 | 551 | 63 | ||
GenBank mRNA | AF076673.1 | - | 1 - 2 | 51 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
